Kobalt 1.000 mg/l Co in dil. salpetersyre (fra Co) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455262UEA 1700 NOK
455262U 455264W
Kobalt 1.000 mg/l Co in dil. salpetersyre (fra Co) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Cobalt Standards
Co in HNO₃ 2 to 5%.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.