Kobber 10.000 mg/l i fortynnet salpetersyre (fra Cu) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455294FEA 7000 NOK
455294F 455292D
Kobber 10.000 mg/l i fortynnet salpetersyre (fra Cu) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Copper Standards
Cu in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.