Dysprosium 1.000 mg/l i fortynnet salpetersyre (fra Dy2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455302LEA 825 NOK
455302L 455304N
Dysprosium 1.000 mg/l i fortynnet salpetersyre (fra Dy2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Dysprosium Standards
Dy₂O₃ in 2 to 5% HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.