Erbium 1.000 mg/l i fortynnet salpetersyre (fra Er2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455322PEA 550 NOK
455322P 455324R
Erbium 1.000 mg/l i fortynnet salpetersyre (fra Er2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Erbium Standards
Er₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.