Europium 1.000 mg/l i fortynnet salpetersyre (fra Eu2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455342TEA 3050 NOK
455342T 455344V
Europium 1.000 mg/l i fortynnet salpetersyre (fra Eu2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Europium Standards
Eu₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.