Gadolinium 1.000 mg/l i fortynnet salpetersyre (de Gd2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455362AEA 2625 NOK
455362A 455364C
Gadolinium 1.000 mg/l i fortynnet salpetersyre (de Gd2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Gadolinium Standards
Gd₂O₃ in HNO₃ (2 to 5%).
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.