Gallium 1.000 mg/l i fortynnet salpetersyre (fra Ga) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455382EEA 2600 NOK
455382E 455384G
Gallium 1.000 mg/l i fortynnet salpetersyre (fra Ga) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Gallium Standards
Ga in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.