Holmium 1.000 mg/l i fortynnet salpetersyre (de Ho2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455462DEA 3050 NOK
455462D 455464F
Holmium 1.000 mg/l i fortynnet salpetersyre (de Ho2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Holmium Standards
Ho₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.