Indium 1.000 mg/l i fortynnet salpetersyre (fra In) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455484JEA 5775 NOK
455484J 455482H
Indium 1.000 mg/l i fortynnet salpetersyre (fra In) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Indium Standards
In in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.