Iridium 1.000 mg/l i 10% saltsyre (fra IrCl3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455502REA 4150 NOK
455502R 455504T
Iridium 1.000 mg/l i 10% saltsyre (fra IrCl3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Iridium Standards
IrCl₃.3H₂O in HCl 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.