Lantan 1.000 mg/l i fortynnet salpetersyre (fra La2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455542CEA 1395 NOK
455542C 455544E
Lantan 1.000 mg/l i fortynnet salpetersyre (fra La2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Lanthanum Standards
La₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.