Litium 10.000 mg/l i fortynnet salpetersyre (fra Li2CO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455592MEA 835 NOK
455592M 455594X
Litium 10.000 mg/l i fortynnet salpetersyre (fra Li2CO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Lithium Standards
Li₂CO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.