Lutetium 1.000 mg/l i fortynnet salpetersyre (fra Lu2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455602UEA 6325 NOK
455602U 455604W
Lutetium 1.000 mg/l i fortynnet salpetersyre (fra Lu2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Lutetium Standards
Lu₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.