Magnesium 1.000 mg/l i fortynnet salpetersyre (fra MgO) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455622BEA 1550 NOK
455622B 455624D
Magnesium 1.000 mg/l i fortynnet salpetersyre (fra MgO) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Magnesium Standards
MgO in HNO₃ 2 to 5%.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.