Molybden 10.000 mg/l Mo in ammonium hydroxide solution (max. 1%) (fra (NH4)2MoO4) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455692PEA 3475 NOK
455692P
Molybden 10.000 mg/l Mo in ammonium hydroxide solution (max. 1%) (fra (NH4)2MoO4) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Molybdenum Standards
(NH₄)₂MoO₄ in HNO₃ tr. HF.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.