Neodymium 1.000 mg/l i fortynnet salpetersyre (fra Nd2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455702AEA 2850 NOK
455702A 455704C
Neodymium 1.000 mg/l i fortynnet salpetersyre (fra Nd2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Neodymium Standards
Nd₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.