Neodymium 10.000 mg/l i fortynnet salpetersyre (fra Nd2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455712CEA 5450 NOK
455712C 455714E
Neodymium 10.000 mg/l i fortynnet salpetersyre (fra Nd2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Neodymium Standards
Nd₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.