Nikkel 10.000 mg/l i fortynnet salpetersyre (fra Ni) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455732GEA 3475 NOK
455732G 455734Y
Nikkel 10.000 mg/l i fortynnet salpetersyre (fra Ni) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Nickel Standards
Ni in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.