Fosfor 1.000 mg/l i vann (fra (NH4)PO4H2) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455784SEA 3300 NOK
455784S
Fosfor 1.000 mg/l i vann (fra (NH4)PO4H2) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Phosphorus Standards
NH₄H₂PO₄ in H₂O.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.