Kalium 1.000 mg/l i fortynnet salpetersyre (fra KNO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455822HEA 1065 NOK
455822H 455824J
Kalium 1.000 mg/l i fortynnet salpetersyre (fra KNO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Potassium Standards
KNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Limited Time Sale
Select items below are currently on sale