Praseodym 1.000 mg/l i fortynnet salpetersyre (fra Pr6O11) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455842LEA 2725 NOK
455842L 455844N
Praseodym 1.000 mg/l i fortynnet salpetersyre (fra Pr6O11) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Praseodymium Standards
Pr₆O₁₁ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.