Rubidium 1.000 mg/l i fortynnet salpetersyre (fra RbNO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455902GEA 2600 NOK
455902G
Rubidium 1.000 mg/l i fortynnet salpetersyre (fra RbNO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Rubidium Standards
RbNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with Certificate of Analysis.