Scandium 10.000 mg/l i fortynnet salpetersyre (fra Sc2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455972UEA 9400 NOK
455972U 455974W
Scandium 10.000 mg/l i fortynnet salpetersyre (fra Sc2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Scandium Standards
Sc₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.