Selen 1.000 mg/l i fortynnet salpetersyre (fra Se) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
455982WEA 1065 NOK
455982W 455984B
Selen 1.000 mg/l i fortynnet salpetersyre (fra Se) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Selenium Standards
Se in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.