Sølv 10.000 mg/l i fortynnet salpetersyre (fra AgNO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456032MEA 3525 NOK
456032M
Sølv 10.000 mg/l i fortynnet salpetersyre (fra AgNO3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Silver Standards
AgNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.