Natrium, standardløsninger for ICP, ARISTAR®
Leverandør: VWR Chemicals
456042XEA 1050 NOK
456042X 456052Q 456054S 456044Q
Natrium, standardløsninger for ICP, ARISTAR®
Standarder Kationstandarder Sodium Standards
NaNO₃ in HNO₃ 2 to 5%.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.