1-Amino-4-bromoanthraquinone-2-sulphonic acid sodium salt, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: 1-Amino-4-bromo-9,10-dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt, 1-Amino-4-bromoanthraquinone-2-sulphonic Acid Sodium Salt, Bromaminic acid Sodium Salt, Bromaminic acid sodium salt, Sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate

M-Clarity™ quality level = MQ100

456063-25G
456063-25GEA 846 NOK
456063-25G
1-Amino-4-bromoanthraquinone-2-sulphonic acid sodium salt, Sigma-Aldrich®
1-Amino-4-bromoanthraquinone-2-sulphonic acid sodium salt
Formel: C₁₄H₈BrNO₅S
MW: 404.17 g/mol
Smeltepunkt: >300 °C (lit.)
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00019160
CAS-nummer: 6258-06-6
EINECS: 228-391-5

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Orange to Red
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Titration with AgNO₃ After Oxygen Combustion Specification: Report Results
Purity (HPLC) ≥83.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR