Strontium 10.000 mg/l i fortynnet salpetersyre (fra Sr(NO3)2) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456072UEA 3525 NOK
456072U 456074W
Strontium 10.000 mg/l i fortynnet salpetersyre (fra Sr(NO3)2) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Strontium Standards
Sr(NO₃)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.