Svovel 1.000 mg/l i vann (fra (NH4)2SO4) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456082WEA 1675 NOK
456082W 456084B
Svovel 1.000 mg/l i vann (fra (NH4)2SO4) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Sulphur Standards
(NH₄)₂SO₄ in H₂O.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.