Terbium 1.000 mg/l i fortynnet salpetersyre (fra Tb4O7) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456142REA 1920 NOK
456142R 456144T
Terbium 1.000 mg/l i fortynnet salpetersyre (fra Tb4O7) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Terbium Standards
Tb₄O₇ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.