VWR® ARISTAR®, Terbium Standard Solution, 10000 mg/L Tb in dil. Nitric Acid (from Tb₄O₇), Standard for ICP
Supplier: VWR Chemicals
456152TEA 6400 NOK
456152T 456154V
VWR® ARISTAR®, Terbium Standard Solution, 10000 mg/L Tb in dil. Nitric Acid (from Tb₄O₇), Standard for ICP
Standards Cationic Standards Terbium Standards
Tb₄O₇ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Certifications: Supplied with certificate of analysis.
Order Now