Thallium 1.000 mg/l i fortynnet salpetersyre (fra Tl) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456162VEA 1775 NOK
456162V
Thallium 1.000 mg/l i fortynnet salpetersyre (fra Tl) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Thallium Standards
Tl in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.