Tinn 10.000 mg/l i 10% saltsyre (fra Sn) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456252WEA 3550 NOK
456252W
Tinn 10.000 mg/l i 10% saltsyre (fra Sn) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Tin Standards
Sn in HCl.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.