Vanadium 1.000 mg/l i fortynnet salpetersyre (fra V2O5) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456322TEA 1750 NOK
456322T 456324V
Vanadium 1.000 mg/l i fortynnet salpetersyre (fra V2O5) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Vanadium Standards
V₂O₅ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.