Ytterbium 1.000 mg/l i fortynnet salpetersyre (fra Yb2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456344CEA 2175 NOK
456344C
Ytterbium 1.000 mg/l i fortynnet salpetersyre (fra Yb2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Ytterbium Standards
Yb₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with Certificate of Analysis.