Yttrium 1.000 mg/l i fortynnet salpetersyre (fra Y2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456362EEA 1065 NOK
456362E 456364G
Yttrium 1.000 mg/l i fortynnet salpetersyre (fra Y2O3) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Yttrium Standards
Y₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.