Zink 1.000 mg/l i fortynnet salpetersyre (fra Zn) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456384KEA 2055 NOK
456384K 456382Y
Zink 1.000 mg/l i fortynnet salpetersyre (fra Zn) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Zinc Standards
Zn in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.