Zirkonium 1.000 mg/l i fortynnet salpetersyre (fra ZrO(NO3)2) ARISTAR® enkeltgrunnstoffsstandard til ICP
Leverandør: VWR Chemicals
456402SEA 2040 NOK
456402S 456404U
Zirkonium 1.000 mg/l i fortynnet salpetersyre (fra ZrO(NO3)2) ARISTAR® enkeltgrunnstoffsstandard til ICP
Standarder Kationstandarder Zirconium Standards
ZrO(NO₃)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.