VWR® ARISTAR®, ICP Multi-Element Calibration Standard 2 in dilute Nitric Acid
Leverandør: VWR Chemicals
456462HEA 0 NOK
456462H
VWR® ARISTAR®, ICP Multi-Element Calibration Standard 2 in dilute Nitric Acid
Standarder Kationstandarder Multi-Element Standards
In 2% nitric acid solution.
3 elements: 250 mg/L Li; 1000 mg/L Na; 10000 mg/L K
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with Certificate of Analysis.