VWR® ARISTAR®, ICP Multi-Element Calibration Standard 3 in dilute Nitric Acid
Leverandør: VWR Chemicals
456472JEA 6125 NOK
456472J
VWR® ARISTAR®, ICP Multi-Element Calibration Standard 3 in dilute Nitric Acid
Standarder Kationstandarder Multi-Element Standards
In 2% nitric acid solution.
4 Elements: 1000 mg/L Ba, Ca, Mg, Sr
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.