Salpetersyre 5%, ARISTAR® Calibration_blank_ICP
Leverandør: VWR Chemicals
Danger
Synonymer: Nitric acid solution
456484NEA 1550 NOK
456484N
Salpetersyre 5%, ARISTAR® Calibration_blank_ICP
Salpetersyre
Calibration blank for ICP, 5% HNO₃ in ASTM Type II Water
Sertifiseringer: Tested in ISO Guide 34 /ISO17025 accredited laboratory.
Bestillingsinformasjon: Supplied with certificate of analysis.
Leveringsinformasjon: Supplied with certificate of analysis.
Formel: HNO₃
MW: 63.01 g/mol
Kokepunkt: 87
Smeltepunkt: -42
Tetthet: 1.51
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011349
CAS-nummer: 7697-37-2
EINECS: 231-714-2
FN: 2031
ADR: 8 II
Merck Indeks: 13,06610