Saltsyre 5%, ARISTAR® Calibration_blank_ICP

Leverandør: VWR Chemicals
Danger

Synonymer: Hydrochloric acid solution, Muriatic acid

456494PEA 1500 NOK
456494P
Saltsyre 5%, ARISTAR® Calibration_blank_ICP
Saltsyre

5% HCl in ASTM Type II Water


Bestillingsinformasjon: Supplied with certificate of analysis.

Formel: HCl
MW: 36.46 g/mol
MDL nummer: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
FN: 1789
ADR: 8 III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR