VWR® ARISTAR®, Multi-Element Calibration Standard 2 in dil. Nitric Acid for ICP-MS
Leverandør: VWR Chemicals
456502VEA 8275 NOK
456502V
VWR® ARISTAR®, Multi-Element Calibration Standard 2 in dil. Nitric Acid for ICP-MS
Standarder Kationstandarder Multi-Element Standards
29 Elements in 5% nitric acid solution.
29 Elements 10 mg/L : Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.