VWR® ARISTAR®, Multi-Element Calibration Standard 3 in Hydrochloric Acid 10% for ICP-MS
Leverandør: VWR Chemicals
456512AEA 4325 NOK
456512A
VWR® ARISTAR®, Multi-Element Calibration Standard 3 in Hydrochloric Acid 10% for ICP-MS
Standarder Kationstandarder Multi-Element Standards
10 Elements in 10% Hydrochloric acid solution.
10 Elements 10 mg/l : Au, Hf, Ir, Pd, Pt, Rh, Ru, Sb, Sn, Te
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.