VWR®, Tuning solution 1 in dil. nitric acid ARISTAR® for ICP-MS
Leverandør: VWR Chemicals
456532EEA 5450 NOK
456532E
VWR®, Tuning solution 1 in dil. nitric acid ARISTAR® for ICP-MS
Standarder Kationstandarder Multi-Element Standards
8 Components in 2% HNO₃.
8 Elements 100 mg/L : Ba, Be, Cu, In, Li, Mg, Tl, U
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.