VWR® ARISTAR®, Tuning Solution 2 in dil. Nitric Acid for ICP-MS
Leverandør: VWR Chemicals
456542GEA 8850 NOK
456542G
VWR® ARISTAR®, Tuning Solution 2 in dil. Nitric Acid for ICP-MS
Standarder Kationstandarder Multi-Element Standards
13 Components in 2% HNO₃.
13 components 100 mg/L : Ba, Be, Bi, Ce, Cu, Ho, In, Li, Mg, Pb, Tl, U, Y
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.