VWR® ARISTAR®, Interference Check A in Nitric Acid 1% for ICP-MS
Leverandør: VWR Chemicals
456552YEA 12375 NOK
456552Y
VWR® ARISTAR®, Interference Check A in Nitric Acid 1% for ICP-MS
Standarder Kationstandarder Multi-Element Standards
12 Elements in 1% nitric acid solution.
12 Elements:18000 mg/L Cl; 1000 mg/L Al, K, Mg, P, S; 2000 mg/L C; 3000 mg/L Ca; 2500 mg/L Fe, Na; 20 mg/L Mo, Ti
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.