VWR® ARISTAR®, Multi-Element Quality Control Standard 2 in dil. Nitric Acid for ICP-MS
Leverandør: VWR Chemicals
456602BEA 5685 NOK
456602B
VWR® ARISTAR®, Multi-Element Quality Control Standard 2 in dil. Nitric Acid for ICP-MS
Standarder Kationstandarder Multi-Element Standards
25 Elements in 5% nitric acid solution.
25 Elements 10 mg/L : Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.