VWR® ARISTAR®, Multi-Element Calibration Standard 1 in dil. Nitric Acid for ICP-MS
Leverandør: VWR Chemicals
456622FEA 6125 NOK
456622F
VWR® ARISTAR®, Multi-Element Calibration Standard 1 in dil. Nitric Acid for ICP-MS
Standarder Kationstandarder Multi-Element Standards
17 Elements in 5% nitric acid solution.
17 Elements 10 mg/ml : Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Sertifiseringer: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformasjon: Supplied with certificate of analysis.