VWR® ARISTAR®, Manganese Standard Solutions for ICP-MS
Leverandør: VWR Chemicals
456802HEA 1925 NOK
456802H 85579.180
VWR® ARISTAR®, Manganese Standard Solutions for ICP-MS
Standarder Kationstandarder Manganese Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0,5% (typical)
  • Delivered with complete certificate of analysis
Sertifiseringer: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities